Assay Details
| Gene Symbol: LOC4336813 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780245.2 | XP_015635731.1 | 2 - 3 | 306 | 142 | |
| GenBank mRNA | AK061833.1 | - | 2 - 3 | 299 | 142 | |
| AK098964.1 | - | 2 - 3 | 295 | 142 | ||
| AK100053.1 | - | 2 - 3 | 300 | 142 | ||
More Information
Set Membership: |
|

LOC4336813
Genomic Map