Assay Details
| Gene Symbol: LOC4336813 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780245.2 | XP_015635731.1 | 6 - 7 | 895 | 103 | |
| GenBank mRNA | AK061833.1 | - | 6 - 7 | 889 | 103 | |
| AK098964.1 | - | 6 - 7 | 884 | 103 | ||
| AK100053.1 | - | 6 - 7 | 889 | 103 | ||
More Information
Set Membership: |
|

LOC4336813
Genomic Map