Assay Details
| Gene Symbol: LOC4336917 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781094.2 | XP_015636580.1 | 4 - 5 | 670 | 108 | |
| GenBank mRNA | AK060852.1 | - | 2 - 3 | 173 | 108 | |
| AK102588.1 | - | 4 - 5 | 648 | 108 | ||
More Information
Set Membership: |
|

LOC4336917
Genomic Map