Assay Details
| Gene Symbol: LOC4336917 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781094.2 | XP_015636580.1 | 9 - 10 | 1196 | 97 | |
| GenBank mRNA | AK060852.1 | - | 7 - 8 | 699 | 97 | |
| AK102588.1 | - | 9 - 10 | 1174 | 97 | ||
More Information
Set Membership: |
|

LOC4336917
Genomic Map