Assay Details
| Gene Symbol: LOC4336983 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781223.1 | XP_015636709.1 | 11 - 12 | 1074 | 108 | |
| GenBank mRNA | AK061967.1 | - | 7 - 8 | 492 | 108 | |
| AK102306.1 | - | 11 - 12 | 1075 | 108 | ||
More Information
Set Membership: |
|

LOC4336983
Genomic Map