Assay Details
| Gene Symbol: LOC4336983 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781223.1 | XP_015636709.1 | 13 - 14 | 1204 | 65 | |
| GenBank mRNA | AF251073.1 | - | 3 - 4 | 168 | 65 | |
| AK061967.1 | - | 9 - 10 | 622 | 65 | ||
| AK102306.1 | - | 13 - 14 | 1205 | 65 | ||
More Information
Set Membership: |
|

LOC4336983
Genomic Map