Assay Details
Gene Symbol: LOC4337137 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015780183.2 | XP_015635669.1 | 18 - 19 | 1813 | 131 | |
GenBank mRNA | AK070546.1 | - | 18 - 19 | 1803 | 131 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
