Assay Details
Gene Symbol: LOC4337137 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015780183.2 | XP_015635669.1 | 2 - 3 | 315 | 75 | |
GenBank mRNA | AK070546.1 | - | 2 - 3 | 305 | 75 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015780183.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
