Assay Details
| Gene Symbol: LOC4337313 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781342.2 | XP_015636828.1 | 3 - 4 | 1202 | 142 | |
| GenBank mRNA | AK243269.1 | - | 3 - 4 | 993 | 142 | |
| AY578162.1 | - | 4 - 5 | 869 | 142 | ||
More Information
Set Membership: |
|

LOC4337313
Genomic Map