Assay Details
| Gene Symbol: LOC4337313 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781342.2 | XP_015636828.1 | 4 - 5 | 1613 | 71 | |
| GenBank mRNA | AK243269.1 | - | 4 - 5 | 1404 | 71 | |
| AY578162.1 | - | 5 - 6 | 1280 | 71 | ||
More Information
Set Membership: |
|

LOC4337313
Genomic Map