Assay Details
Gene Symbol: LOC4337737 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK101387.1 | - | 3 - 4 | 863 | 80 |
Gene Symbol: LOC9272524 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK287884.1 | - | 1747 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
