Assay Details
Gene Symbol: LOC4337737 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015781898.2 | XP_015637384.1 | 2 - 3 | 575 | 72 | |
XM_015781899.2 | XP_015637385.1 | 4 - 5 | 1031 | 72 | ||
XM_015781900.2 | XP_015637386.1 | 4 - 5 | 1013 | 72 | ||
XM_015781901.2 | XP_015637387.1 | 2 - 3 | 575 | 72 | ||
XM_015781902.2 | XP_015637388.1 | 2 - 3 | 575 | 72 | ||
GenBank mRNA | AK101387.1 | - | 4 - 5 | 958 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
