Assay Details
Gene Symbol: LOC4337785 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015784663.2 | XP_015640149.1 | 4 - 5 | 941 | 95 | |
XM_015784664.1 | XP_015640150.1 | 4 - 5 | 570 | 95 | ||
GenBank mRNA | AK120211.1 | - | 4 - 5 | 577 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
