Assay Details
Gene Symbol: LOC4338744 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015781814.2 | XP_015637300.1 | 1 - 2 | 412 | 75 | |
GenBank mRNA | AK071730.1 | - | 1 - 2 | 395 | 75 | |
AK099324.1 | - | 1 - 2 | 319 | 75 | ||
AK104169.1 | - | 1 - 2 | 390 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
