Assay Details
Gene Symbol: LOC4339019 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015782432.2 | XP_015637918.1 | 4 - 5 | 357 | 64 | |
XM_015782433.2 | XP_015637919.1 | 4 - 5 | 354 | 64 | ||
GenBank mRNA | AK058842.1 | - | 4 - 5 | 337 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
