Assay Details
| Gene Symbol: LOC4339019 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015782432.2 | XP_015637918.1 | 7 - 8 | 534 | 110 | |
| XM_015782433.2 | XP_015637919.1 | 7 - 8 | 531 | 110 | ||
| GenBank mRNA | AK058842.1 | - | 7 - 8 | 514 | 110 | |
More Information
Set Membership: |
|

LOC4339019
Genomic Map