Assay Details
Gene Symbol: LOC4339019 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015782432.2 | XP_015637918.1 | 8 - 9 | 638 | 67 | |
XM_015782433.2 | XP_015637919.1 | 8 - 9 | 635 | 67 | ||
GenBank mRNA | AK058842.1 | - | 8 - 9 | 618 | 67 | |
AK066350.1 | - | 5 - 6 | 2119 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
