Assay Details
Gene Symbol: LOC4339406 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015782182.2 | XP_015637668.1 | 4 - 5 | 753 | 64 | |
XM_015782183.2 | XP_015637669.1 | 4 - 5 | 750 | 64 | ||
GenBank mRNA | AK108715.1 | - | 4 - 5 | 706 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
