Assay Details
| Gene Symbol: LOC4339406 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015782182.2 | XP_015637668.1 | 6 - 7 | 878 | 150 | |
| XM_015782183.2 | XP_015637669.1 | 6 - 7 | 875 | 150 | ||
| GenBank mRNA | AK064168.1 | - | 6 - 7 | 836 | 150 | |
| AK108715.1 | - | 6 - 7 | 831 | 150 | ||
More Information
Set Membership: |
|

LOC4339406
Genomic Map