Assay Details
Gene Symbol: LOC4339406 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015782183.2 | XP_015637669.1 | 8 - 9 | 1012 | 118 | |
GenBank mRNA | AK062298.1 | - | 8 - 9 | 982 | 118 | |
AK108715.1 | - | 8 - 9 | 968 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
