Assay Details
Gene Symbol: LOC4339406 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015782182.2 | XP_015637668.1 | 9 - 10 | 1162 | 63 | |
XM_015782183.2 | XP_015637669.1 | 9 - 10 | 1141 | 63 | ||
GenBank mRNA | AK108715.1 | - | 9 - 10 | 1097 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
