Assay Details
Gene Symbol: LOC4339457 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015783720.2 | XP_015639206.1 | 8 - 9 | 1250 | 73 | |
XM_015783721.1 | XP_015639207.1 | 7 - 8 | 1178 | 73 | ||
XM_015783722.2 | XP_015639208.1 | 8 - 9 | 1202 | 73 | ||
GenBank mRNA | AK068572.1 | - | 8 - 9 | 971 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
