Assay Details
| Gene Symbol: LOC4339810 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784662.1 | XP_015640148.1 | 1 - 2 | 177 | 114 | |
| GenBank mRNA | AK121070.1 | - | 1 - 2 | 154 | 114 | |
| BT131017.1 | - | 1 - 2 | 132 | 114 | ||
| BT131438.2 | - | 1 - 2 | 152 | 114 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015784662.1 transcripts.
Set Membership: |
|

LOC4339810
Genomic Map