Assay Details
| Gene Symbol: LOC4340192 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785450.1 | XP_015640936.1 | 5 - 6 | 702 | 75 | |
| GenBank mRNA | AK059986.1 | - | 5 - 6 | 702 | 75 | |
| AK101378.1 | - | 5 - 6 | 703 | 75 | ||
More Information
Set Membership: |
|

LOC4340192
Genomic Map