Assay Details
Gene Symbol: LOC9268973 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015787609.2 | XP_015643095.1 | 26 - 27 | 3551 | 115 | |
GenBank mRNA | AK065390.1 | - | 6 - 7 | 529 | 115 |

More Information
Set Membership: |
![]() ![]() ![]() |
