Assay Details
Gene Symbol: LOC9268973 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015787609.2 | XP_015643095.1 | 28 - 29 | 3828 | 144 | |
GenBank mRNA | AK065390.1 | - | 8 - 9 | 806 | 144 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
