Assay Details
Gene Symbol: LOC4340386 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015787539.2 | XP_015643025.1 | 10 - 11 | 1453 | 106 | |
XM_015787540.2 | XP_015643026.1 | 10 - 11 | 1420 | 106 | ||
XM_015787541.2 | XP_015643027.1 | 10 - 11 | 2062 | 106 | ||
GenBank mRNA | AK072074.1 | - | 10 - 11 | 1374 | 106 | |
AK098923.1 | - | 10 - 11 | 1337 | 106 | ||
AK100546.1 | - | 10 - 11 | 2068 | 106 | ||
AK119193.1 | - | 6 - 7 | 810 | 106 | ||
EF122480.1 | - | 10 - 11 | 1319 | 106 | ||
EF122481.1 | - | 10 - 11 | 1354 | 106 | ||
HQ895720.1 | - | 9 - 10 | 1287 | 106 | ||
JF969237.1 | - | 9 - 10 | 1306 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
