Assay Details
Gene Symbol: LOC4340386 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015787539.2 | XP_015643025.1 | 14 - 15 | 2410 | 60 | |
XM_015787540.2 | XP_015643026.1 | 14 - 15 | 2377 | 60 | ||
XM_015787541.2 | XP_015643027.1 | 14 - 15 | 3019 | 60 | ||
GenBank mRNA | AK072074.1 | - | 14 - 15 | 2331 | 60 | |
AK098923.1 | - | 14 - 15 | 2294 | 60 | ||
AK100546.1 | - | 14 - 15 | 3025 | 60 | ||
AK119193.1 | - | 10 - 11 | 1767 | 60 | ||
EF122480.1 | - | 14 - 15 | 2276 | 60 | ||
EF122481.1 | - | 14 - 15 | 2311 | 60 | ||
HQ895720.1 | - | 13 - 14 | 2244 | 60 | ||
JF969237.1 | - | 13 - 14 | 2263 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
