Assay Details
Gene Symbol: LOC4340386 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015787539.2 | XP_015643025.1 | 1 - 2 | 156 | 64 | |
GenBank mRNA | AK072074.1 | - | 1 - 2 | 77 | 64 | |
AK098923.1 | - | 1 - 2 | 40 | 64 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015787539.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
