Assay Details
Gene Symbol: LOC4340386 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015787539.2 | XP_015643025.1 | 4 - 5 | 539 | 86 | |
XM_015787540.2 | XP_015643026.1 | 4 - 5 | 506 | 86 | ||
XM_015787541.2 | XP_015643027.1 | 4 - 5 | 1148 | 86 | ||
GenBank mRNA | AK072074.1 | - | 4 - 5 | 460 | 86 | |
AK098923.1 | - | 4 - 5 | 423 | 86 | ||
AK100546.1 | - | 4 - 5 | 1154 | 86 | ||
EF122480.1 | - | 4 - 5 | 405 | 86 | ||
EF122481.1 | - | 4 - 5 | 86 | |||
HQ895720.1 | - | 3 - 4 | 373 | 86 | ||
JF969237.1 | - | 3 - 4 | 392 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
