Assay Details
Gene Symbol: LOC4340386 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015787539.2 | XP_015643025.1 | 5 - 6 | 729 | 70 | |
XM_015787540.2 | XP_015643026.1 | 5 - 6 | 696 | 70 | ||
XM_015787541.2 | XP_015643027.1 | 5 - 6 | 1338 | 70 | ||
GenBank mRNA | AK072074.1 | - | 5 - 6 | 650 | 70 | |
AK098923.1 | - | 5 - 6 | 613 | 70 | ||
AK100546.1 | - | 5 - 6 | 1344 | 70 | ||
AK119193.1 | - | 1 - 2 | 86 | 70 | ||
EF122480.1 | - | 5 - 6 | 595 | 70 | ||
EF122481.1 | - | 5 - 6 | 630 | 70 | ||
HQ895720.1 | - | 4 - 5 | 563 | 70 | ||
JF969237.1 | - | 4 - 5 | 582 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
