Assay Details
Gene Symbol: LOC4340386 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015787539.2 | XP_015643025.1 | 9 - 10 | 1339 | 68 | |
XM_015787540.2 | XP_015643026.1 | 9 - 10 | 1306 | 68 | ||
XM_015787541.2 | XP_015643027.1 | 9 - 10 | 1948 | 68 | ||
GenBank mRNA | AK072074.1 | - | 9 - 10 | 1260 | 68 | |
AK098923.1 | - | 9 - 10 | 1223 | 68 | ||
AK100546.1 | - | 9 - 10 | 1954 | 68 | ||
AK119193.1 | - | 5 - 6 | 696 | 68 | ||
EF122480.1 | - | 9 - 10 | 1205 | 68 | ||
EF122481.1 | - | 9 - 10 | 1240 | 68 | ||
HQ895720.1 | - | 8 - 9 | 1173 | 68 | ||
JF969237.1 | - | 8 - 9 | 1192 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
