Assay Details
Gene Symbol: LOC4330897 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015771783.2 | XP_015627269.1 | 16 - 17 | 2035 | 116 | |
GenBank mRNA | AK068672.1 | - | 16 - 17 | 1801 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
