Assay Details
Gene Symbol: LOC4330897 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015771783.2 | XP_015627269.1 | 22 - 23 | 2824 | 67 | |
GenBank mRNA | AK068672.1 | - | 22 - 23 | 2590 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
