Assay Details
| Gene Symbol: LOC4341470 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK059035.1 | - | 1 - 2 | 350 | 113 | |
| AK070190.1 | - | 2 - 3 | 1376 | 113 | ||
| AK072098.1 | - | 2 - 3 | 1376 | 113 | ||
More Information
Set Membership: |
|

LOC4341470
Genomic Map