Assay Details
| Gene Symbol: LOC4341500 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788540.2 | XP_015644026.1 | 2 - 3 | 458 | 138 | |
| GenBank mRNA | AK072533.1 | - | 2 - 3 | 366 | 138 | |
| AK099872.1 | - | 2 - 3 | 364 | 138 | ||
More Information
Set Membership: |
|

LOC4341500
Genomic Map