Assay Details
| Gene Symbol: LOC4341500 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788540.2 | XP_015644026.1 | 3 - 4 | 657 | 86 | |
| GenBank mRNA | AK072533.1 | - | 3 - 4 | 565 | 86 | |
| AK099872.1 | - | 3 - 4 | 86 | |||
More Information
Set Membership: |
|

LOC4341500
Genomic Map