Assay Details
Gene Symbol: LOC4341716 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015786186.1 | XP_015641672.1 | 1 - 2 | 261 | 85 | |
GenBank mRNA | AK060123.1 | - | 1 - 2 | 233 | 85 | |
AK060127.1 | - | 1 - 2 | 226 | 85 | ||
AK104184.1 | - | 1 - 2 | 230 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
