Assay Details
Gene Symbol: LOC9272258 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015787386.2 | XP_015642872.1 | 1 | 644 | 183 | |
GenBank mRNA | AK288023.1 | - | 1 | 619 | 183 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
