Assay Details
Gene Symbol: LOC4342177 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015790797.2 | XP_015646283.1 | 7 - 8 | 991 | 81 | |
GenBank mRNA | AK100327.1 | - | 7 - 8 | 983 | 81 | |
AK100796.1 | - | 7 - 8 | 983 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
