Assay Details
Gene Symbol: LOC4342214 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015789772.2 | XP_015645258.1 | 2 - 3 | 442 | 145 | |
XM_015789773.2 | XP_015645259.1 | 2 - 3 | 442 | 145 | ||
XM_015789775.2 | XP_015645261.1 | 2 - 3 | 442 | 145 | ||
XM_015789776.2 | XP_015645262.1 | 2 - 3 | 442 | 145 | ||
GenBank mRNA | AB003325.1 | - | 2 - 3 | 385 | 145 | |
AY551920.1 | - | 2 - 3 | 428 | 145 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
