Assay Details
| Gene Symbol: LOC4342214 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015789772.2 | XP_015645258.1 | 4 - 5 | 613 | 68 | |
| XM_015789773.2 | XP_015645259.1 | 4 - 5 | 613 | 68 | ||
| XM_015789775.2 | XP_015645261.1 | 4 - 5 | 613 | 68 | ||
| XM_015789776.2 | XP_015645262.1 | 4 - 5 | 613 | 68 | ||
| GenBank mRNA | AB003325.1 | - | 4 - 5 | 556 | 68 | |
| AK072683.1 | - | 3 - 4 | 1948 | 68 | ||
| AY551920.1 | - | 4 - 5 | 599 | 68 | ||
More Information
Set Membership: |
|

LOC4342214
Genomic Map