Assay Details
Gene Symbol: LOC4342542 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015790069.2 | XP_015645555.1 | 11 - 12 | 843 | 74 | |
GenBank mRNA | AK066812.1 | - | 11 - 12 | 792 | 74 | |
AK067073.1 | - | 11 - 12 | 803 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
