Assay Details
Gene Symbol: LOC4343296 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015792215.2 | XP_015647701.1 | 1 - 2 | 154 | 68 | |
XM_015792216.2 | XP_015647702.1 | 1 - 2 | 154 | 68 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015792215.2, XM_015792216.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
