Assay Details
| Gene Symbol: LOC4343612 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791776.2 | XP_015647262.1 | 1 - 2 | 318 | 86 | |
| GenBank mRNA | AK067125.1 | - | 1 - 2 | 91 | 86 | |
| AK067240.1 | - | 1 - 2 | 326 | 86 | ||
| AK068892.1 | - | 1 - 2 | 325 | 86 | ||
| EU847007.1 | - | 1 - 2 | 66 | 86 | ||
More Information
Set Membership: |
|

LOC4343612
Genomic Map