Assay Details
Gene Symbol: LOC9267772 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015789467.2 | XP_015644953.1 | 7 - 8 | 1366 | 85 | |
GenBank mRNA | AK065600.1 | - | 7 - 8 | 1148 | 85 | |
AK103165.1 | - | 7 - 8 | 1148 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
