Assay Details
Gene Symbol: LOC4344253 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015792084.2 | XP_015647570.1 | 1 - 2 | 96 | 142 | |
GenBank mRNA | AK060100.1 | - | 1 - 2 | 61 | 142 | |
AK103678.1 | - | 1 - 2 | 63 | 142 | ||
AK104794.1 | - | 1 - 2 | 61 | 142 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015792084.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
