Assay Details
Gene Symbol: LOC4344253 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015792084.2 | XP_015647570.1 | 2 - 3 | 214 | 61 | |
GenBank mRNA | AK060100.1 | - | 2 - 3 | 179 | 61 | |
AK103678.1 | - | 2 - 3 | 181 | 61 | ||
AK104794.1 | - | 2 - 3 | 179 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
