Assay Details
Gene Symbol: LOC4344253 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015792084.2 | XP_015647570.1 | 3 - 4 | 286 | 115 | |
GenBank mRNA | AK060100.1 | - | 3 - 4 | 251 | 115 | |
AK103678.1 | - | 3 - 4 | 253 | 115 | ||
AK104794.1 | - | 3 - 4 | 251 | 115 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
