Assay Details
Gene Symbol: LOC4344253 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015792084.2 | XP_015647570.1 | 7 - 8 | 746 | 89 | |
GenBank mRNA | AK060100.1 | - | 7 - 8 | 711 | 89 | |
AK103678.1 | - | 7 - 8 | 712 | 89 | ||
AK104794.1 | - | 7 - 8 | 711 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
